Learn More
Cayman Chemical 1 2DIlInoleoyl3Oleoylrac 10mg
SDP
Catalog No. 502937109
Encompass_Preferred
A triacylglycerol; has been found in ostrich and emu oils, as well as in the fat body of male B. lapidarius bumblebees; reduces scald development on apples of the Delicious variety when applied at 3% w/v immediately following harvest and assessed after six months of storage
Catalog No. 50-293-7109 Supplier Cayman Chemical Supplier No. 2817810 MG
A triacylglycerol; has been found in ostrich and emu oils, as well as in the fat body of male B. lapidarius bumblebees; reduces scald development on apples of the Delicious variety when applied at 3% w/v immediately following harvest and assessed after six months of storage