Learn More
Cayman Chemical 1,2-Dilinoleoyl-3-Palmitoyl-rac-glycerol, Purity: ≥95%, CAS: 2190-15-0, 50 mg
SDP
Catalog No. 502934536
Encompass_Preferred
A triacylglycerol; has been found in a variety of vegetable oils; reduces scald development on apples of the Delicious variety when applied a concentration of 3% w/v immediately following harvest and assessed after six months of storage
Catalog No. 50-293-4536 Supplier Cayman Chemical Supplier No. 2688750 MG
A triacylglycerol; has been found in a variety of vegetable oils; reduces scald development on apples of the Delicious variety when applied a concentration of 3% w/v immediately following harvest and assessed after six months of storage