Learn More
Cayman Chemical 1 2DIlInoleoylsnglycerol 10mg
SDP
Catalog No. 502935026
Encompass_Preferred
A DAG; has been found as a component of phosphatidic acid in rat liver mitochondria and in spinach chloroplast membranes; upregulated in some pregnant women and has been used as a biomarker to predict later preeclampsia in EArly pregnancy
Catalog No. 50-293-5026 Supplier Cayman Chemical Supplier No. 2542210 MG
A DAG; has been found as a component of phosphatidic acid in rat liver mitochondria and in spinach chloroplast membranes; upregulated in some pregnant women and has been used as a biomarker to predict later preeclampsia in EArly pregnancy