Learn More
Cayman Chemical 1,2-Dilinoleoyl-sn-glycerol, Purity: ≥95%, CAS: 24529-89-3, 50 mg
SDP
Catalog No. 502936046
Encompass_Preferred
A DAG; has been found as a component of phosphatidic acid in rat liver mitochondria and in spinach chloroplast membranes; upregulated in some pregnant women and has been used as a biomarker to predict later preeclampsia in early pregnancy
Catalog No. 50-293-6046 Supplier Cayman Chemical Supplier No. 2542250 MG
A DAG; has been found as a component of phosphatidic acid in rat liver mitochondria and in spinach chloroplast membranes; upregulated in some pregnant women and has been used as a biomarker to predict later preeclampsia in early pregnancy