Learn More
Cayman Chemical 1 2DInonadecnoylsnglycero 10mg
SDP
Catalog No. 502936416
Encompass_Preferred
A saturated phospholipid; has been used as a standard for the quantification of phosphatidylcholines in human synovial fluid; has been used to study dynamics of lipid bilayer phase transition
Catalog No. 50-293-6416 Supplier Cayman Chemical Supplier No. 2559710 MG
A saturated phospholipid; has been used as a standard for the quantification of phosphatidylcholines in human synovial fluid; has been used to study dynamics of lipid bilayer phase transition