Learn More
Cayman Chemical 1,2-Dioctanoyl-3-Decanoyl-rac-glycerol, Purity: ≥95%, CAS: 69095-45-0, 10 mg
SDP
Catalog No. 502939489
Encompass_Preferred
A triacylglycerol; has been found in the suberin of wound-healing potatoes
Catalog No. 50-293-9489 Supplier Cayman Chemical Supplier No. 2855410 MG
A triacylglycerol; has been found in the suberin of wound-healing potatoes