Learn More
Cayman Chemical 1,2-Dioleoyl-3(R)-trimethylammoniumpropane (chloride), Purity: ≥95%, CAS: 328250-28-8, 100 mg
SDP
Catalog No. 502915609
Encompass_Preferred
A cationic lipid; lipoplexes containing R-DOTAP and encapsulating siRNA targeting CYP19A1 decrease aromatase activity in MCF-7 cells; vaccination with an HPV 16 protein E7 peptide in R-DOTAP-containing liposomes induces IFN-γ production by CD8+ T cells and TILs and decreases tumor size in a murine cervical cancer model; subcutaneous immunization with recombinant influenza B hemagglutinin using R-DOTAP as an adjuvant increases IL-2 and IFN-γ production in CD4+ T cells isolated from mouse draining lymph nodes and spleen
Catalog No. 50-291-5609 Supplier Cayman Chemical Supplier No. 38194100 MG
A cationic lipid; lipoplexes containing R-DOTAP and encapsulating siRNA targeting CYP19A1 decrease aromatase activity in MCF-7 cells; vaccination with an HPV 16 protein E7 peptide in R-DOTAP-containing liposomes induces IFN-γ production by CD8+ T cells and TILs and decreases tumor size in a murine cervical cancer model; subcutaneous immunization with recombinant influenza B hemagglutinin using R-DOTAP as an adjuvant increases IL-2 and IFN-γ production in CD4+ T cells isolated from mouse draining lymph nodes and spleen