Learn More
Cayman Chemical 1,2-Dioleoyl-3(S)-trimethylammoniumpropane (chloride), Purity: ≥95%, CAS: 428506-51-8, 50 mg
SDP
Catalog No. 502916473
Encompass_Preferred
A cationic lipid; lipoplexes containing S-DOTAP and encapsulating siRNA targeting CYP19A1 decrease aromatase activity in MCF-7 cells; vaccination with an HPV 16 protein E7 peptide in S-DOTAP-containing liposomes decrease tumor size but do not induce IFN-γ production by CD8+ T cells and TILs and in a murine cervical cancer model
Catalog No. 50-291-6473 Supplier Cayman Chemical Supplier No. 3819550 MG
A cationic lipid; lipoplexes containing S-DOTAP and encapsulating siRNA targeting CYP19A1 decrease aromatase activity in MCF-7 cells; vaccination with an HPV 16 protein E7 peptide in S-DOTAP-containing liposomes decrease tumor size but do not induce IFN-γ production by CD8+ T cells and TILs and in a murine cervical cancer model