Learn More
Cayman Chemical 1,2-Dioleoyl-3-Decanoyl-rac-glycerol, Purity: ≥98%, CAS: 88286-47-9, 5 mg
SDP
Catalog No. 502938385
Encompass_Preferred
A triacylglycerol; has been found in date seed oil
Catalog No. 50-293-8385 Supplier Cayman Chemical Supplier No. 269315 MG
A triacylglycerol; has been found in date seed oil