Learn More
Cayman Chemical 1 2DIoleoyl3Docosnoylrac 10mg
SDP
Catalog No. 502940357
Encompass_Preferred
A triacylglycerol; has been found in cocoa butter and in CBEs containing shea stearin and palm mid fraction
Catalog No. 50-294-0357 Supplier Cayman Chemical Supplier No. 2886310 MG
A triacylglycerol; has been found in cocoa butter and in CBEs containing shea stearin and palm mid fraction