Learn More
Cayman Chemical 1,2-Dioleoyl-3-Myristoyl-rac-glycerol, Purity: ≥98%, CAS: 35804-98-9, 5 mg
SDP
Catalog No. 502937685
Encompass_Preferred
A triacylglycerol; has been found in the fat body of male B. lapidarius bumblebees and in R. erythropolis bacteria utilizing phenol or succinic acid as a carbon source
Catalog No. 50-293-7685 Supplier Cayman Chemical Supplier No. 272915 MG
A triacylglycerol; has been found in the fat body of male B. lapidarius bumblebees and in R. erythropolis bacteria utilizing phenol or succinic acid as a carbon source