Learn More
Cayman Chemical 1,2-Dioleoyl-3-trimethylammoniumpropane (methyl sulfate), Purity: ≥98%, CAS: 144189-73-1, 10 mg
SDP
Catalog No. 502919078
Encompass_Preferred
A cationic lipid; has been used for transfection of DNA, RNA, and other negatively charged molecules into eukaryotic cells; has been used in the composition of small unilamellar liposomes formulated as gene delivery vectors for gene therapy
Catalog No. 50-291-9078 Supplier Cayman Chemical Supplier No. 3890510 MG
A cationic lipid; has been used for transfection of DNA, RNA, and other negatively charged molecules into eukaryotic cells; has been used in the composition of small unilamellar liposomes formulated as gene delivery vectors for gene therapy