Learn More
Cayman Chemical 1,2-Dioleoyl-sn-glycero-3-EPC (chloride), Purity: ≥95%, CAS: 474945-24-9, 10 mg
SDP
Catalog No. 502911411
Encompass_Preferred
A cationic esterized phospholipid; has been used in combination with other lipids in the formation of lipid bilayers
Catalog No. 50-291-1411 Supplier Cayman Chemical Supplier No. 3647310 MG
A cationic esterized phospholipid; has been used in combination with other lipids in the formation of lipid bilayers