Learn More
Cayman Chemical 1,2-Dioleoyl-sn-glycero-3-PA (ammonium salt), Purity: ≥95%, 5 mg
SDP
Catalog No. 502984159
Encompass_Preferred
A phospholipid; has been used in the generation of micelles, liposomes, and other artificial membranes
Catalog No. 50-298-4159 Supplier Cayman Chemical Supplier No. 209565 MG
A phospholipid; has been used in the generation of micelles, liposomes, and other artificial membranes