Learn More
Cayman Chemical 1,2-Dioleoyl-sn-glycero-3-PC, Purity: ≥98%, CAS: 4235-95-4, 5 g
SDP
Catalog No. 502970141
Encompass_Preferred
A phospholipid; has commonly been used alone, or with other components, in the generation of micelles, liposomes, and other types of artificial membranes; has also been used in combination with other lipids in the generation of LNPs for the delivery of mRNA and microRNA in vivo; intravenous administration of LNPs containing DOPC and encapsulating luciferase mRNA induce a 4-fold higher expression of luciferase in mouse liver compared to LNPs containing DOPE or DSPC; has been used in the formation of artificial lipid droplets in combination with 1,2,3-trioleoyl glycerol for the study of lipid peroxidation in lipid droplets
Catalog No. 50-297-0141 Supplier Cayman Chemical Supplier No. 150985 G
A phospholipid; has commonly been used alone, or with other components, in the generation of micelles, liposomes, and other types of artificial membranes; has also been used in combination with other lipids in the generation of LNPs for the delivery of mRNA and microRNA in vivo; intravenous administration of LNPs containing DOPC and encapsulating luciferase mRNA induce a 4-fold higher expression of luciferase in mouse liver compared to LNPs containing DOPE or DSPC; has been used in the formation of artificial lipid droplets in combination with 1,2,3-trioleoyl glycerol for the study of lipid peroxidation in lipid droplets