Learn More
Cayman Chemical 1 2DIoleoylsnglycero3PG 10mg
SDP
Catalog No. 502983881
Encompass_Preferred
A phospholipid containing the long-chain (18:1) fatty acid oleic acid inserted at the sn-1 and sn-2 positions
Catalog No. 50-298-3881 Supplier Cayman Chemical Supplier No. 2095710 MG
A phospholipid containing the long-chain (18:1) fatty acid oleic acid inserted at the sn-1 and sn-2 positions