Learn More
Cayman Chemical 1,2-Dioleoyl-sn-glycero-3-PS (sodium salt), Purity: ≥95%, CAS: 90693-88-2, 10 mg
SDP
Catalog No. 502978745
Encompass_Preferred
A phospholipid; SapC-1,2-DOPS nanovesicles are cytotoxic to SK-MEL-28 cells in vitro and decrease tumor volume in a squamous cell carcinoma mouse xenograft model; has been used in the formation of unilamellar vesicles to study the effect of curvature on membrane structure and in the formation of supported lipid bilayers to study the effects of various support materials on lipid redistribution between membrane leaflets
Catalog No. 50-297-8745 Supplier Cayman Chemical Supplier No. 2998310 MG
A phospholipid; SapC-1,2-DOPS nanovesicles are cytotoxic to SK-MEL-28 cells in vitro and decrease tumor volume in a squamous cell carcinoma mouse xenograft model; has been used in the formation of unilamellar vesicles to study the effect of curvature on membrane structure and in the formation of supported lipid bilayers to study the effects of various support materials on lipid redistribution between membrane leaflets