Learn More
Cayman Chemical 1,2-Dipalmitoyl Ethylene Glycol, Purity: ≥95%, CAS: 624-03-3, 100 mg
SDP
Catalog No. 502934313
Encompass_Preferred
A fatty acid ester; has been used in the manufacture of plastics, moldable flame-retardant resin compositions, and as a component of antifouling paint; can migrate out of plastics but is not toxic to rats during prenatal development at 900 mg/kg per day
Catalog No. 50-293-4313 Supplier Cayman Chemical Supplier No. 25590100 MG
A fatty acid ester; has been used in the manufacture of plastics, moldable flame-retardant resin compositions, and as a component of antifouling paint; can migrate out of plastics but is not toxic to rats during prenatal development at 900 mg/kg per day