Learn More
Cayman Chemical 1 2DIpalmItoyl Ethylen Glyc
SDP
Supplier:  Cayman Chemical 25590250 MG
Encompass_Preferred
A fatty acid ester; has been used in the manufacture of plastics, moldable flame-retardant resin compositions, and as a component of antifouling paint; can migrate out of plastics but is not toxic to rats during prenatal development at 900 mg/kg per day
Catalog No. 50-293-4280