Learn More
Cayman Chemical 1 2DIpalmItoyl3ElaIdoylra 10mg
SDP
Catalog No. 502941187
Encompass_Preferred
A triacylglycerol; has been used as a standard for the identification of triacylglycerols in rapeseed and palm oils
Catalog No. 50-294-1187 Supplier Cayman Chemical Supplier No. 2886510 MG
A triacylglycerol; has been used as a standard for the identification of triacylglycerols in rapeseed and palm oils