Learn More
Cayman Chemical 1 2DIpalmItoyl3trImthylam 10mg
SDP
Catalog No. 502918837
Encompass_Preferred
A cationic lipid; has been used as a transfection reagent in vitro and in vivo and in the formation of liposomes and microbubbles; microbubbles containing DPTAP and encapsulating miR-145 reduce atherosclerotic plaque area by 48.04% using UTMD in ApoE-/- mice in a model of atherosclerosis induced by a high-fat diet
Catalog No. 50-291-8837 Supplier Cayman Chemical Supplier No. 3890710 MG
A cationic lipid; has been used as a transfection reagent in vitro and in vivo and in the formation of liposomes and microbubbles; microbubbles containing DPTAP and encapsulating miR-145 reduce atherosclerotic plaque area by 48.04% using UTMD in ApoE-/- mice in a model of atherosclerosis induced by a high-fat diet