Learn More
Cayman Chemical 1,2-Dipalmitoyl-sn-glycero-3-EPC (chloride), Purity: ≥95%, CAS: 328250-18-6, 10 mg
SDP
Catalog No. 502973205
Encompass_Preferred
A phospholipid containing the saturated long-chain (16:0) stearic acid inserted at the sn-1 and sn-2 positions and an alkyl group on the phosphate oxygen of the polar headgroup; may be used as DNA transfection agent and for studies of surface charge density within lipid bilayers
Catalog No. 50-297-3205 Supplier Cayman Chemical Supplier No. 1509910 MG
A phospholipid containing the saturated long-chain (16:0) stearic acid inserted at the sn-1 and sn-2 positions and an alkyl group on the phosphate oxygen of the polar headgroup; may be used as DNA transfection agent and for studies of surface charge density within lipid bilayers