Learn More
Cayman Chemical 1,2-Dipalmitoyl-sn-glycero-3-PE-N-(cap biotin) (sodium salt), Purity: ≥95%, CAS: 384835-52-3, 10 mg
SDP
Catalog No. 502936481
Encompass_Preferred
A biotinylated phospholipid; has been used in various SLB systems to isolate circulating tumor cells and tumor cell microembolis from patient-derived blood by antibody-coated microfluidics, detect protein-ligand binding, or create DNA curtains in nanowells for high-throughput detection of protein-DNA interactions at the single molecule level
Catalog No. 50-293-6481 Supplier Cayman Chemical Supplier No. 2572410 MG
A biotinylated phospholipid; has been used in various SLB systems to isolate circulating tumor cells and tumor cell microembolis from patient-derived blood by antibody-coated microfluidics, detect protein-ligand binding, or create DNA curtains in nanowells for high-throughput detection of protein-DNA interactions at the single molecule level