Learn More
Cayman Chemical 1 2DIphytnoylsnglycero3 10mg
SDP
Catalog No. 502970139
Encompass_Preferred
A phospholipid containing the tetramethylated long-chain (16:0) diphytanic acid inserted at the sn-1 and sn-2 positions
Catalog No. 50-297-0139 Supplier Cayman Chemical Supplier No. 1509410 MG
A phospholipid containing the tetramethylated long-chain (16:0) diphytanic acid inserted at the sn-1 and sn-2 positions