Learn More
Cayman Chemical 1,2-Diphytanoyl-sn-glycero-3-PC, Purity: ≥98%, CAS: 207131-40-6, 10 mg
SDP
Catalog No. 502936379
Encompass_Preferred
A phospholipid; has been used in the creation of lipid bilayer membranes; has been used, in combination with LPS, to form the outer leaflet of a tethered bilayer lipid membrane as a model of a Gram-negative bacterial membrane
Catalog No. 50-293-6379 Supplier Cayman Chemical Supplier No. 2559810 MG
A phospholipid; has been used in the creation of lipid bilayer membranes; has been used, in combination with LPS, to form the outer leaflet of a tethered bilayer lipid membrane as a model of a Gram-negative bacterial membrane