Learn More
Cayman Chemical 1,2-Distearoyl-3-Arachidoyl-rac-glycerol, Purity: ≥98%, CAS: 77145-69-8, 10 mg
SDP
Catalog No. 502941116
Encompass_Preferred
A triacylglycerol; has been found in butterfat
Catalog No. 50-294-1116 Supplier Cayman Chemical Supplier No. 2738610 MG
A triacylglycerol; has been found in butterfat