Learn More
Cayman Chemical 1 2DIstearoyl3PalmItoylra 10mg
SDP
Catalog No. 502941781
Encompass_Preferred
A triacylglycerol; has been found in cocoa butter, lard, and cod liver oil
Catalog No. 50-294-1781 Supplier Cayman Chemical Supplier No. 2855110 MG
A triacylglycerol; has been found in cocoa butter, lard, and cod liver oil