Learn More
Cayman Chemical 1,2-Distearoyl-3-Palmitoyl-rac-glycerol, Purity: ≥95%, CAS: 2177-99-3, 5 mg
SDP
Catalog No. 502941778
Encompass_Preferred
A triacylglycerol; has been found in cocoa butter, lard, and cod liver oil
Catalog No. 50-294-1778 Supplier Cayman Chemical Supplier No. 285515 MG
A triacylglycerol; has been found in cocoa butter, lard, and cod liver oil