Learn More
Cayman Chemical 1 2DIstearoyld70snglycero 10mg
SDP
Catalog No. 502939186
Encompass_Preferred
An internal standard for the quantification of 1,2-distearoyl-sn-glycero-3-PC by GC- or LC-MS
Catalog No. 50-293-9186 Supplier Cayman Chemical Supplier No. 2875210 MG
An internal standard for the quantification of 1,2-distearoyl-sn-glycero-3-PC by GC- or LC-MS