Learn More
Cayman Chemical 1,2-Distearoyl-sn-glycero-3-PC (solution), Purity: ≥98%, CAS: 816-94-4, 10 mL
SDP
Catalog No. 502915723
Encompass_Preferred
A phospholipid; has commonly been used in the generation of micelles, liposomes, and other types of artificial membranes
Catalog No. 50-291-5723 Supplier Cayman Chemical Supplier No. 3918910 ML
A phospholipid; has commonly been used in the generation of micelles, liposomes, and other types of artificial membranes