Learn More
LGC GENOMICS LLC 1/2G Molecular Trap Pack, Maintains Anhydrous Conditions, Removes Water and Contaminants, 0.5 g
Catalog No. NC1995811
Encompass
The LGC Biosearch Technologies 1/2G Molecular Trap Pack is designed to effectively remove water and small particulate contaminants from solvents and solutions, ensuring anhydrous conditions for sensitive applications like oligonucleotide synthesis and NMR. Its highly activated molecular sieves maintain water levels below 50 ppm, preventing reagent degradation and instrument clogging. The tightly sealed packaging enhances its effectiveness and reliability, making it a crucial tool in laboratory environments.
Catalog No. NC1995811 Supplier LGC GENOMICS LLC Supplier No. BG713901
The LGC Biosearch Technologies 1/2G Molecular Trap Pack is designed to effectively remove water and small particulate contaminants from solvents and solutions, ensuring anhydrous conditions for sensitive applications like oligonucleotide synthesis and NMR. Its highly activated molecular sieves maintain water levels below 50 ppm, preventing reagent degradation and instrument clogging. The tightly sealed packaging enhances its effectiveness and reliability, making it a crucial tool in laboratory environments.