Learn More
Pipeline Packaging Corp 1/2G ROUND CAN UNLINED EA
SDP
Catalog No. NC3735258
Encompass_Preferred
1/2G ROUND CAN UNLINED EA.American-made 1/2 Gallon Tinplate Round Paint Can, a superior packaging solution that comes unlined, with a case count of 114, and is characterized by its high-quality construction and design, ensuring optimal protection and preservation of its contents. With a height of 5 7/8" and a diameter of 5 3/8", this metal can is expertly crafted to meet the versatile needs of various industries such as paint, chemical, food, and more.
Catalog No. NC3735258 Supplier Pipeline Packaging Corp Supplier No. 40050EA
1/2G ROUND CAN UNLINED EA.American-made 1/2 Gallon Tinplate Round Paint Can, a superior packaging solution that comes unlined, with a case count of 114, and is characterized by its high-quality construction and design, ensuring optimal protection and preservation of its contents. With a height of 5 7/8" and a diameter of 5 3/8", this metal can is expertly crafted to meet the versatile needs of various industries such as paint, chemical, food, and more.