Learn More
Ace Glass, Inc. 1/2gal high temperature silicone based bath oil
SDP
Catalog No. 509559122
Encompass
An extremely stable, medium viscosity, silicone oil. Temperature range High Temp. maximum 230 degree C. Oil shows no discoloration after 40 hours at maximum temperature. Qty 0.5 Gallon
Catalog No. 50-955-9122 Supplier Ace Glass, Inc. Supplier No. 1411514
An extremely stable, medium viscosity, silicone oil. Temperature range High Temp. maximum 230 degree C. Oil shows no discoloration after 40 hours at maximum temperature. Qty 0.5 Gallon