Learn More
Ace Glass, Inc. 1/2gal low temp bath oil silicone base, 180c.
SDP
Catalog No. 509559120
Encompass
An extremely stable, medium viscosity, silicone oil. Temperature range: Low Temp. maximum 180 degree COil shows no discoloration after 40 hours at maximum temperature.
Catalog No. 50-955-9120 Supplier Ace Glass, Inc. Supplier No. 1411510
An extremely stable, medium viscosity, silicone oil. Temperature range: Low Temp. maximum 180 degree COil shows no discoloration after 40 hours at maximum temperature.