Learn More
Grainger BWAY Pail, 1/2 gal
Catalog No. NC1929827
Encompass_Preferred
The BWAY 1/2 gallon plastic pail is a round container made of high density polyethylene and has a diameter of 6-1/2 inches. It features a bail handle for easy carrying and a screw-on lid with a gasket for a tight seal. The pail has a capacity of 64 fluid ounces. It can be used for storing and transporting a variety of materials including food, chemicals, and industrial supplies. The pail is FDA-approved and made in the USA. With its durable construction and leakproof lid, this BWAY pail is ideal for commercial and industrial use.
Catalog No. NC1929827 Supplier Grainger Supplier No. 7J110
The BWAY 1/2 gallon plastic pail is a round container made of high density polyethylene and has a diameter of 6-1/2 inches. It features a bail handle for easy carrying and a screw-on lid with a gasket for a tight seal. The pail has a capacity of 64 fluid ounces. It can be used for storing and transporting a variety of materials including food, chemicals, and industrial supplies. The pail is FDA-approved and made in the USA. With its durable construction and leakproof lid, this BWAY pail is ideal for commercial and industrial use.