Learn More
Medchemexpress LLC 1(2H)-Isoquinolinone | 491-30-5 | 145.16 | 25 G
SDP
Catalog No. 5000448044
Encompass_Preferred
1-Hydroxyisoquinoline is a biochemical reagent suitable for life science related research. It can be utilized as a biological material or an organic compound. This product is for research use only and not intended for patient use.
  • Can inhibit DNA repair Poly (ADP-ribose) polymerase 1
  • Inhibits in permeabilized L1210 murine leukemia cells with an IC50 of 15.8 μM
  • Suitable for various life science related research applications
Catalog No. 50-004-48044 Supplier Medchemexpress LLC Supplier No. 25GHYW002014
1-Hydroxyisoquinoline is a biochemical reagent suitable for life science related research. It can be utilized as a biological material or an organic compound. This product is for research use only and not intended for patient use.
  • Can inhibit DNA repair Poly (ADP-ribose) polymerase 1
  • Inhibits in permeabilized L1210 murine leukemia cells with an IC50 of 15.8 μM
  • Suitable for various life science related research applications