Learn More
Medchemexpress LLC 1(2H)-Isoquinolinone | 491-30-5 | MFCD00006899 | 145.16 | 10 G
SDP
Catalog No. 5000447084
Encompass_Preferred
1-Hydroxyisoquinoline is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is intended for research use only.
  • Appearance: Solid, light brown to gray
  • Inhibits DNA repair Poly (ADP-ribose) polymerase 1 in permeabilised L1210 murine leukemia cells
Catalog No. 50-004-47084 Supplier Medchemexpress LLC Supplier No. HYW00201410G
1-Hydroxyisoquinoline is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is intended for research use only.
  • Appearance: Solid, light brown to gray
  • Inhibits DNA repair Poly (ADP-ribose) polymerase 1 in permeabilised L1210 murine leukemia cells