Learn More
Medchemexpress LLC 1(2H)-Isoquinolinone, 5-hydroxy- | 5154-02-9 | MFCD00006900 | 99.1% | 161.16 | C9H7NO2 | 100 MG
SDP
Catalog No. 5000302662
Encompass_Preferred
1,5-Isoquinolinediol is a small-molecule PARP inhibitor (CAS 5154-02-9) used in biochemical and cellular research. It has a reported IC50 of 0.18-0.37 μM and is supplied as a high-purity solid suitable for studies of PARP-mediated pathways and oxidative stress.
  • Potent PARP inhibitor with reported IC50 0.18-0.37 μM.
  • High purity for analytical and biological assays.
  • Available in multiple laboratory-scale sizes, including 100 mg.
  • Supplied with analytical characterization and a certificate of analysis.
  • Commonly used in research on oxidative stress and PARP pathways.
Catalog No. 50-003-02662 Supplier Medchemexpress LLC Supplier No. HYW015422100MG
1,5-Isoquinolinediol is a small-molecule PARP inhibitor (CAS 5154-02-9) used in biochemical and cellular research. It has a reported IC50 of 0.18-0.37 μM and is supplied as a high-purity solid suitable for studies of PARP-mediated pathways and oxidative stress.
  • Potent PARP inhibitor with reported IC50 0.18-0.37 μM.
  • High purity for analytical and biological assays.
  • Available in multiple laboratory-scale sizes, including 100 mg.
  • Supplied with analytical characterization and a certificate of analysis.
  • Commonly used in research on oxidative stress and PARP pathways.