Learn More
Medchemexpress LLC 1(2H)-isoquinolinone, 5-hydroxy- | 5154-02-9 | MFCD00006900 | 99.1% | 161.16 g·mol⁻¹ | C9H7NO2 | 1 ML
SDP
Catalog No. 5000303753
Encompass_Preferred
1,5-Isoquinolinediol is a small-molecule poly(ADP-ribose) polymerase (PARP) inhibitor (CAS 5154-02-9) used in biochemical research. It exhibits in vitro potency with reported IC50 values of 0.18-0.37 μM and is employed to study PARP-mediated oxidative stress, including diabetes-induced NADPH oxidase-derived retinal damage.
  • High purity suitable for research applications.
  • Reported IC50 0.18-0.37 μM against PARP.
  • Molecular formula C9H7NO2; molecular weight 161.16 g·mol⁻¹.
  • Available as solid and as a 10 mM solution in DMSO.
  • Used to study PARP-related oxidative stress and retinal models.
  • Supplied with a certificate of analysis for batch-specific quality.
Catalog No. 50-003-03753 Supplier Medchemexpress LLC Supplier No. HYW01542210MM/1ML
1,5-Isoquinolinediol is a small-molecule poly(ADP-ribose) polymerase (PARP) inhibitor (CAS 5154-02-9) used in biochemical research. It exhibits in vitro potency with reported IC50 values of 0.18-0.37 μM and is employed to study PARP-mediated oxidative stress, including diabetes-induced NADPH oxidase-derived retinal damage.
  • High purity suitable for research applications.
  • Reported IC50 0.18-0.37 μM against PARP.
  • Molecular formula C9H7NO2; molecular weight 161.16 g·mol⁻¹.
  • Available as solid and as a 10 mM solution in DMSO.
  • Used to study PARP-related oxidative stress and retinal models.
  • Supplied with a certificate of analysis for batch-specific quality.