Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1(2H)-Naphthalenone, 6-bromo-3,4-dihydro- | 66361-67-9 | 225.09 | 5 G
SDP

Catalog No. 5000447415
Encompass_Preferred

6-Bromo-3,4-dihydro-1(2H)-naphthalenone is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is intended for research use only and not for sale to patients.

  • Biochemical reagent for life science related research
  • Appearance: off-white to pink solid
  • Chemical formula: C10H9BrO
  • Soluble in DMSO
  • For research use only
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-47415 Supplier Medchemexpress LLC Supplier No. HYW0083215G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Bromo-3,4-dihydro-1(2H)-naphthalenone is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is intended for research use only and not for sale to patients.

  • Biochemical reagent for life science related research
  • Appearance: off-white to pink solid
  • Chemical formula: C10H9BrO
  • Soluble in DMSO
  • For research use only
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.