Learn More
MSC Value Collection Machine Screw, 8-32, 1/2 in, Pan Phillips, 100/Pk.
Catalog No. NC1895604
Encompass
These machine screws are constructed from grade 18-8 stainless steel, designed for reliable fastening in various applications. They feature a pan head and Phillips drive, providing secure installation and corrosion resistance.
  • Thread size: #8-32
  • Length: 1/2 in, measured under head
  • Head type: Pan
  • Drive type: Phillips
  • Material grade: 18-8 stainless steel
  • Finish: Bright/uncoated
Catalog No. NC1895604 Supplier MSC Supplier No. 67571521
These machine screws are constructed from grade 18-8 stainless steel, designed for reliable fastening in various applications. They feature a pan head and Phillips drive, providing secure installation and corrosion resistance.
  • Thread size: #8-32
  • Length: 1/2 in, measured under head
  • Head type: Pan
  • Drive type: Phillips
  • Material grade: 18-8 stainless steel
  • Finish: Bright/uncoated