Learn More
Roboz Surgical 1/2IN RND BURS 0.6MM 5PK
SDP
Catalog No. NC9775404
Encompass
Carbide Round Burs; Size #1/2, 0.6mm Tips; 1.75 in. Length; 2.33mm Shaft Diameter; Pack Of 5 For use with the RS-6300 Ideal Micro Drill
Catalog No. NC9775404 Supplier Roboz Surgical Supplier No. RS6280C1/2
Carbide Round Burs; Size #1/2, 0.6mm Tips; 1.75 in. Length; 2.33mm Shaft Diameter; Pack Of 5 For use with the RS-6300 Ideal Micro Drill