Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Tube VAC 1/2 ID x 1 OD x 8 ft

Catalog No. NC9673808 Shop All Thermo Scientific Products
Encompass_Preferred

Tube VAC 1/2 ID x 1 OD x 8 ft.

Catalog No. NC9673808 Supplier Thermo Scientific Supplier No. M30000703
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tube VAC 1/2 ID x 1 OD x 8 ft.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.