Learn More
Cole-Parmer BriskHeat TBIH051-020LD Dual Element, Cloth Insulated Heat Tape, 2' x 1/2" , 156 Watts, 120 V
Catalog No. NC9181761 Shop All Cole-Parmer Products
Encompass_Preferred
Dual-element design improves the heaters flexibility over standard high-temperature tapes. Tapes are made of two heating elements that are knitted together in series and then wound together with high-temperature Samox fiberglass. The 120 VAC tapes include a plug Warning: Heating tapes should not be overlapped and require the use of a temperature or power controller sold separately.
Catalog No. NC9181761 Supplier Cole-Parmer Supplier No. 3605000
Dual-element design improves the heaters flexibility over standard high-temperature tapes. Tapes are made of two heating elements that are knitted together in series and then wound together with high-temperature Samox fiberglass. The 120 VAC tapes include a plug Warning: Heating tapes should not be overlapped and require the use of a temperature or power controller sold separately.