Learn More
Chemglass Life Sciences Adapter, TFE Compression, 1/2" OD to 3/4" NPT Male
SDP
Catalog No. 501531354
Encompass_Preferred
Adapter, TFE Compression, 1/2" OD to 3/4" NPT Male. Designed for use with Chemglass's FEP tubing. Fittings are made entirely of chemically inert corrosion resistant PTFE. Fittings: Machined from virgin Polytetrafluoroethylene. Ferrules: Ferrules are machined from virgin Polytetrafluoroethylene. The Gripper in the two piece Ferrule-Gripper assemblies are manufactured from PVDF. Nuts are molded PVDF.
Catalog No. 50-153-1354 Supplier Chemglass Life Sciences Supplier No. CG197181K
Adapter, TFE Compression, 1/2" OD to 3/4" NPT Male. Designed for use with Chemglass's FEP tubing. Fittings are made entirely of chemically inert corrosion resistant PTFE. Fittings: Machined from virgin Polytetrafluoroethylene. Ferrules: Ferrules are machined from virgin Polytetrafluoroethylene. The Gripper in the two piece Ferrule-Gripper assemblies are manufactured from PVDF. Nuts are molded PVDF.