Learn More
Thomas Scientific 1.2mL 96-Well Plate, Clear, Round Well, U-Bottom, Shared Wall, 50 case
SDP
Catalog No. NC1757609
Encompass
Ideal for assay work, magnetic beads, and sample storage. The unique open bottom design offers more area for magnetic beads, while larger diameter wells improve sample access and mixing. Raised rims on each well make for easy and secure sealing with adhesive films, silicone mats, or heat sealing films. Manufactured from virgin polypropylene and certified to be free of RNase, DNase, and Pyrogen contamination.
Catalog No. NC1757609 Supplier Thomas Scientific Supplier No. 1149J39
Add to Cart
Add to Cart
Ideal for assay work, magnetic beads, and sample storage. The unique open bottom design offers more area for magnetic beads, while larger diameter wells improve sample access and mixing. Raised rims on each well make for easy and secure sealing with adhesive films, silicone mats, or heat sealing films. Manufactured from virgin polypropylene and certified to be free of RNase, DNase, and Pyrogen contamination.