Learn More
Labcon North America 1.2ML 96WELL PLTS STRL 100/CS
Catalog No. NC2281604
Encompass_Preferred
LABCON 1.2ML STERILE 96 DEEP WELL PLATES WITH SQUARE/V BOTTOMThese square hole flat bottom plates hold a full 2.5 mL** and work as storage plates or reservoirs.
This item is not returnable. View return policy
Catalog No. NC2281604 Supplier Labcon North America Supplier No. 39085250009
LABCON 1.2ML STERILE 96 DEEP WELL PLATES WITH SQUARE/V BOTTOMThese square hole flat bottom plates hold a full 2.5 mL** and work as storage plates or reservoirs.