Learn More
Chemglass Life Sciences Chemglass 1.2 mL Cryogenic Vial, Self-Standing, Internal Thread, Sterile (10 x 10 per rack; 1,400/case)
SDP
Catalog No. 502733346
Encompass_Preferred
Sterile 1.2 mL self-standing cryogenic vials with internal threads designed for secure sample storage at cryogenic temperatures. Manufactured from USP 6 grade raw materials in a controlled clean-room environment, the vials are E-beam sterilized, non-pyrogenic, and DNase/RNase free. Supplied racked in nests for easy handling and inventory management.
  • Capacity: 1.2 mL
  • Internal thread
  • Self-standing, round bottom design
  • Sterile (E-beam sterilized)
  • Manufactured from USP 6 grade raw materials
  • Pack/rack quantities: 10 x 10 per rack; 14 racks per case (1,400 vials/case)
  • Autoclavable; recommended storage in the gas phase of a nitrogen tank
  • Non-pyrogenic; DNase/RNase free
  • Manufactured in a 100,000 grade clean room; ISO 9001 and ISO 13485 certified
Catalog No. 50-273-3346 Supplier Chemglass Life Sciences Supplier No. CGN4766007
Sterile 1.2 mL self-standing cryogenic vials with internal threads designed for secure sample storage at cryogenic temperatures. Manufactured from USP 6 grade raw materials in a controlled clean-room environment, the vials are E-beam sterilized, non-pyrogenic, and DNase/RNase free. Supplied racked in nests for easy handling and inventory management.
  • Capacity: 1.2 mL
  • Internal thread
  • Self-standing, round bottom design
  • Sterile (E-beam sterilized)
  • Manufactured from USP 6 grade raw materials
  • Pack/rack quantities: 10 x 10 per rack; 14 racks per case (1,400 vials/case)
  • Autoclavable; recommended storage in the gas phase of a nitrogen tank
  • Non-pyrogenic; DNase/RNase free
  • Manufactured in a 100,000 grade clean room; ISO 9001 and ISO 13485 certified