Learn More
Chemglass Life Sciences Chemglass 1.2 mL Cryogenic Vial, Self-Standing, Internal Thread, Sterile, Racked/Nested (CGN-4766-010)
SDP
Catalog No. 502733343
Encompass_Preferred
A sterile 1.2 mL cryogenic vial with a self-standing round bottom and internal thread, supplied racked and nested for automated handling. Vials are sterilized by e-beam and produced from USP 6 grade materials to meet cleanroom production standards.
  • Capacity 1.2 mL.
  • Self-standing, round bottom vial.
  • Internal thread closure.
  • Sterile, sterilized by e-beam.
  • Packaged 9 x 9 per rack; 14 racks per case; 1,134 vials per case.
  • Manufactured from USP 6 grade raw materials.
  • Produced in a 100,000 grade cleaning room.
  • ISO 9001 and ISO 13485 certified.
  • Solid one-piece construction prevents barcode loss at low temperatures.
  • Clear graduations and white marking area.
  • Autoclavable.
  • Non-pyrogenic and DNase/RNase free.
  • Recommended for storage in the gas phase of a nitrogen tank.
Catalog No. 50-273-3343 Supplier Chemglass Life Sciences Supplier No. CGN4766010
A sterile 1.2 mL cryogenic vial with a self-standing round bottom and internal thread, supplied racked and nested for automated handling. Vials are sterilized by e-beam and produced from USP 6 grade materials to meet cleanroom production standards.
  • Capacity 1.2 mL.
  • Self-standing, round bottom vial.
  • Internal thread closure.
  • Sterile, sterilized by e-beam.
  • Packaged 9 x 9 per rack; 14 racks per case; 1,134 vials per case.
  • Manufactured from USP 6 grade raw materials.
  • Produced in a 100,000 grade cleaning room.
  • ISO 9001 and ISO 13485 certified.
  • Solid one-piece construction prevents barcode loss at low temperatures.
  • Clear graduations and white marking area.
  • Autoclavable.
  • Non-pyrogenic and DNase/RNase free.
  • Recommended for storage in the gas phase of a nitrogen tank.