Learn More
Microsolv Technology Corporation 1.2Ml Mrq Screwtop Max Rec Rsa
Catalog No. 5001452324
Encompass_Preferred
Vials, Screw Top, Glass. Clear, 1.2ml, MRQ™, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. RSA Brand. 100/PK.
Catalog No. 50-014-52324 Supplier Microsolv Technology Corporation Supplier No. 9512S0CVTRS
Vials, Screw Top, Glass. Clear, 1.2ml, MRQ™, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. RSA Brand. 100/PK.